Eppendorf™ Repeater™ stream and Xstream Pipetter Available on GSA/VA Contract for Federal Government customers only.
Combine easy handling with a broad range of features
Manufacturer: Eppendorf™ 4986000831
 View more versions of this product
Catalog No. 13-683-506
  Check Availability
Add to Cart
    Specifications
    Product Type With charging adapter and bundle with charger stand,Pipet with charge stand
    Model Repeater Xstream™
    Battery Type Lithium Ion
    Compatible Tips Eppendorf™ Combitips™ Plus tips
    Finger Style Ergonomic
    For Use With (Equipment) Eppendorf™ Combitips™ Plus tips
    Includes Battery
    Warranty 2 Years
    Number of Channels 1,1
    Volume (Metric) Range 1μL to 50mL
    Volume (Metric) 1μL to 50mL
    Capacity Range 1μL to 50mL
    Compatibility Eppendorf™ Combitips™ Plus tips
  • Description & Specifications
Related Products
Product Content Feedback