Eppendorf™ Research Manifolds for Multichannel Pipetters
Manufacturer: Eppendorf™ 022453921
 View more versions of this product
Catalog No. 13-688-508
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Autoclavable Yes
    For Use With (Equipment) 30 to 300μL Research™ multichannel pipette
    Includes 4 Figure volume display
    Number of Channels 8,8
    Product Type Multichannel Pipette Manifold
  • Description & Specifications
Related Products
Product Content Feedback