Learn More
Eppendorf™ Research Series Adjustable Volume Pipetters
Designed for high performance, reliability and user comfort
Manufacturer:  Eppendorf™ 022471902
 View more versions of this product
Catalog No. 05-402-86
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
±1%, ±1.5%, ±2.5%
Certificate of conformity, EN ISO8655
Digital
20μL Pipette tip
3 Years
Ultra micro
0.5 to 10μL
Yes
20μL Pipette tip
Ergonomic
0.01μL
Gray
Micro
0.5 to 10μL
Accessories