Learn More
Eppendorf™ Research Series Adjustable Volume Pipetters
Designed for high performance, reliability and user comfort
Manufacturer:  Eppendorf™ 022472003
 View more versions of this product
Catalog No. 05-402-88
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Micro
10 to 100μL
±0.8%, ±1%, ±1%
Certificate of conformity, EN ISO8655
Digital
200μL and 300μL pipette tip
3 Years
Micro
10 to 100μL
Yes
200μL and 300μL pipette tip
Ergonomic
0.1μL
Yellow
Accessories