Eppendorf™ Research Series Adjustable Volume Pipetters

Designed for high performance, reliability and user comfort

Manufacturer: Eppendorf™ 022472101

 View more versions of this product

Catalog No. 05-402-90

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Accuracy ±0.6%, ±1%, ±3%
    Autoclavable Yes
    Certifications/Compliance Certificate of conformity, EN ISO8655
    Compatible Tips 1000μL Pipette tip
    Display Digital
    Finger Style Ergonomic
    For Use With (Equipment) 1000μL Pipette tip
    Increments 1μL
    Warranty 3 Years
    Color Blue,Blue
    Model Micro
    Product Type Micro
    Volume (Metric) Range 100 to 1000μL
    Volume (Metric) 100 to 1000μL
    Capacity Range 100 to 1000μL
    Compatibility 1000μL Pipette tip
    Type Micro

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit