Eppendorf™ Series 2000 Adjustable-Volume Pipetters

Compact design includes a single, multifunctional button that controls both pipetting and tip ejection

Manufacturer: Eppendorf™ 022470353

 View more versions of this product

Catalog No. 05-402-51

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Equipment) 2500μL Tip
    Accuracy ±1.5%, ±0.8%, ±0.6%
    Autoclavable Yes
    Certifications/Compliance Certificate of conformity
    Compatible Tips 2500μL Tip
    Finger Style Ergonomic
    Increments 2μL
    UV Monitoring Yes
    Warranty 3 Years
    Color Red,Red
    Model Ultra micro
    Volume (Metric) 500 to 2500μL
    Compatibility 2500μL Tip

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit