Eppendorf™ Series 2000 Fixed-Volume Pipetters
Compact design includes a single, multifunctional button that controls both pipetting and tip ejection
Manufacturer: Eppendorf™ 022470906
 View more versions of this product
Catalog No. 05-402-61
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Accuracy ±0.8%
    Autoclavable Yes
    Certifications/Compliance Individual certificate of conformity
    Compatible Tips Universal fit pipette tip
    For Use With (Equipment) Universal fit pipette tip
    Includes Yellow operating button
    UV Monitoring Yes
    Warranty 3 Years
    Model Fixed and adjustable volume
    Volume (Metric) 40μL
    Compatibility Universal fit pipette tip
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback