Eppendorf™ Snap-Cap Microcentrifuge Flex-Tube™ Tubes

Choose from six colors, including amber for light-sensitive samples

Manufacturer: Eppendorf™ 022363778

 View more versions of this product

Catalog No. 05-402-97

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Microcentrifuge Tube
    Color Clear
    Material Polypropylene
    Includes Flex lid
    Packaging 500/Pk.
    Capacity (Metric) 1.5mL
    Diameter (Metric) Outer 10mm
    Length (Metric) 38mm
    Max. Force 25000xG
    Sterile No
    For Use With (Application) For centrifuging or storing samples
    Cap Style Snap Cap
    Graduated Yes
    Shape Conical
    Closure Type Snap
    Autoclavable Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit