Learn More
Eppendorf™ Snap-Cap Microcentrifuge Safe-Lock™ Tubes Available on GSA/VA Contract for Federal Government customers only.
Polypropylene tube with locking cap
Manufacturer:  Eppendorf™ 925000060
 View more versions of this product
Catalog No. 13-864-252
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Polypropylene
Yes
Natural
42mm
25000xG
−122.8° to 212°F
Hinged Snap Cap
2mL
11mm
Polypropylene
Yes
−86° to 100°C