Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Thermomixer™ R

Click to view available options
Thermomixer R; Without Block; 115V, 50/60Hz
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Requires

Thermoblock (sold separately), choice of four:

  • 05-400-202 for 24 × 0.5mL tubes
  • 05-400-203 for 24 × 1.5mL tubes
  • 05-400-204 for 24 × 2.0mL tubes
  • 05-400-207 for one microplate

Heats, shakes and cools samples all in one unit

Speeds reactions by combining multiple sample preparation steps.

Features

  • 10 different exchangeable thermoblocks
  • Automatic identification and display of maximum revolutions
  • Processes up to 24 microtubes at once
  • Digital display shows both both set and actual parameters
  • Simple controls
  • Adjustable temperature from 13°C below ambient to 99°C
  • Variable mixing speed from 300 to 1400rpm (up to 1500rpm with 0.5mL tubes)
  • Programmable for intermittent mixing with timed rest phases—ideal for enzyme reactions and DNA hybridizations
  • Short-mix function supports quick vortex applications
  • Store parameters for common applications
  • RS-232-C printer interface

Order Info

Dimensions: 9.75L × 8.75W × 4.75 in.H (25 × 22 × 12cm)
Net weight: 7 lb. (3.2kg)

Voltage 120 V
Diameter (Metric) Orbit 0.3 cm
Temperature Range (Metric) Ambient -13°C to 99°C
Display Digital
Electrical Requirements 120 V, 50/60 Hz
Frequency 50/60 Hz
Height (English) 4.9 in.
Height (Metric) 12.5 cm
Temperature Accuracy ±0.5 From 20 to 45, ±2 below 20 and above 45°C
Timer 1 min. to 99:59 Hours
Warranty 2 Years
Wattage 90 w
Weight (English) 7 lb.
Weight (Metric) 3.2 kg
Width (English) 8.7 in.
Width (Metric) 22 cm
Show More Show Less