Eppendorf™ Thermomixer™ R

Heats, shakes and cools samples all in one unit

Manufacturer: Eppendorf™ 022670158

 View more versions of this product

Catalog No. 05-400-206

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


Required Accessories : Thermoblock (sold separately), choice of four:<ul><li>05-400-202 for 24 &times; 0.5mL tubes</li><li>05-400-203 for 24 &times; 1.5mL tubes</li><li>05-400-204 for 24 &times; 2.0mL tubes</li><li>05-400-207 for one microplate</li></ul>

    Specifications

    Temperature Range (Metric) 13 Below ambient to 99°C
    Temperature Accuracy ±0.5 From 20 to 45, ±2 below 20 and above 45°C
    Voltage 230V
    Height (English) 4.9 in.
    Height (Metric) 12.5cm
    Width (English) 8.7 in.
    Width (Metric) 22cm
    Wattage 90w
    Hertz 50/60Hz
    Electrical Requirements 230V 50/60Hz
    Diameter (Metric) Orbit 0.3cm
    Display Digital
    Automation Interface RS232C
    Timer 1 min. to 99:59 Hours
    Weight (English) 7 lb.
    Weight (Metric) 3.2kg
    Warranty 2 Years

    Speeds reactions by combining multiple sample preparation steps.

    Features

    • 10 different exchangeable thermoblocks
    • Automatic identification and display of maximum revolutions
    • Processes up to 24 microtubes at once
    • Digital display shows both both set and actual parameters
    • Simple controls
    • Adjustable temperature from 13°C below ambient to 99°C
    • Variable mixing speed from 300 to 1400rpm (up to 1500rpm with 0.5mL tubes)
    • Programmable for intermittent mixing with timed rest phases—ideal for enzyme reactions and DNA hybridizations
    • Short-mix function supports quick vortex applications
    • Store parameters for common applications
    • RS-232-C printer interface

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit