Learn More
Eppendorf™ Tips 10-1000µL
Filter tips with a two-phase filter for contamination protection. The two filter layers made of flexible, hydrophobic material, fit perfectly in the tip cone and retain practically 100% of all aerosols and biomolecules.
Manufacturer:  Eppendorf™ 925000022
 View more versions of this product
Catalog No. 13-684-240
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »