Learn More
Eppendorf™ Tips 10-1000µL
Filter tips with a two-phase filter for contamination protection. The two filter layers made of flexible, hydrophobic material, fit perfectly in the tip cone and retain practically 100% of all aerosols and biomolecules.
Manufacturer:  Eppendorf™ 925000010
 View more versions of this product
Catalog No. 13-684-234
  Check Availability
Add to cart
Specifications
Description
  • Two filter layers provide dual protection against aerosols and biomolecules to guard your pipette and sample
  • Free of PCR inhibiting additives provide for reliable downstream results
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
  • Lot-specific certificates are available at: www.eppendorf.com/certificates
Description & Specifications
Specifications
Dualfilter Tip
1000μL