Eppendorf™ Titermate™ Multichannel Pipetters: Manifolds
Manufacturer: Eppendorf™ 022454251
 View more versions of this product
Catalog No. 13-688-175
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Accuracy ±1.5 to 0.6%
    Autoclavable Yes
    Includes Individual seals and Ceramic pistons
    Number of Channels 12,12
    Volume (Metric) Range 30 to 300μL
    Product Type Titermate™ multichannel pipette manifold
    Volume (Metric) 30 to 300μL
  • Description & Specifications
Related Products
Product Content Feedback