Learn More
Eppendorf Top Buret
Extremely robust with the display and control elements guaranteeing simple, fatigue-free operation
$1499.00
Specifications
For Use With (Equipment) | Rapid titration |
---|---|
Includes | Recirculation valve, valve toggle, telescopic filling tube, adjustable discharge tube, three adapters 40/38/32mm bottle threads, 2 x 1.5V microbattery, quality certificate |
Volume (Metric) | 25 mL |
Catalog Number | Mfr. No. | Type | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Volume (Metric) | Price | Quantity & Availability | ||||
13-687-273
![]() |
Eppendorf
4965000025 |
50 mL |
Each for $1,499.00
|
|
13-687-272
|
Eppendorf
4965000017 |
25 mL |
Each for $1,499.00
|
|
Description
Extremely robust with the display and control elements guaranteeing simple, fatigue-free operation
The innovative Top Buret from Eppendorf sets new standards for manual titration. A new, pulse-free dispensing technique allows the desired volumes to be dispensed continuously, thus eliminating the need for new liquid aspiration each time. This makes titration simpler and safer, with precision values well within required limits
- Continuous, pulse-free dispensing technique for rapid titration
- Patented safety valve for deairing without loss of reagent
- Voltage supply with long-life primary cells and charging display
- Dispensing range from 0.01mL to 999.9mL
- Removable electronics unit
- Single channel with simple calibration program
- Discharge tube rotatable by 360°
- Adjustable discharge tube: horizontal: 142 to 220mm, vertical: 10 to 200mm
- Telescopic aspiration tube for automatically setting the length from 210mm to 370mm
- Connection for dry tube
- The medium is dispensed by simply rotating the wheels
Model M
- Dispensing rate of 2,500µL per rotation
- Dispensing rate of 5,000µL per rotation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Rapid titration | |
Recirculation valve, valve toggle, telescopic filling tube, adjustable discharge tube, three adapters 40/38/32mm bottle threads, 2 x 1.5V microbattery, quality certificate | |
25 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.