Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Eppendorf Tubes™ Conical Tubes Available on GSA/VA Contract for Federal Government customers only.

Use these tubes for the simple and safe processing of sample volumes up to 5.0mL. Eppendorf Tubes™ represent the perfect option for working with medium–sized sample volumes.

Manufacturer:  Eppendorf™ 0030119460

 View more versions of this product

Catalog No. 14-282-301


Add to cart

Description

Description

  • Conical bottom allows usage in existing adapters and racks
  • Made from transparent polypropylene for easy visibility of pellets
  • Manufactured without use of slip agents, plasticizers and biocides and do not interfere with products
  • Sample evaporation during storage and incubation is minimized by precise lid sealing
  • Accidental lid opening from -86° to 80°C prevented by hinged lids
  • Centrifugation stability of up to 25,000 x g prevents tube breakage
  • Available in batch-certified Eppendorf, PCR clean, Sterile and Biopur™ purity grades
  • Available in Protein LoBind and DNA/RNA LoBind material for maximum recovery of valuable samples

  • Standard (Eppendorf) Quality = non-sterile
  • PCR Clean = non-sterile
  • Sterile= sterile
  • Biopur = sterile

Cell culture, Isolation of plasmid DNA and total RNA, Liquid handling

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Non-sterile
25,000 x g
Clear
5mL
200
Conical
Microcentrifuge Tube, PCR clean
Yes
RNase, DNase-free
0.169 oz.
PP (Polypropylene)
Bag
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit