Learn More
Eppendorf™ twin.tec™ Real-Time 96-Well PCR Platesc Available on GSA/VA Contract for Federal Government customers only.
Fluorescence intensity up to ten times higher than with frosted wells
Manufacturer:  Eppendorf™ 0030132742
 View more versions of this product
Catalog No. 14-281-212
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
Blue
Polypropylene
Skirted
White
Yes
RT-PCR applications
Yes
Yes