Eppendorf™ twin.tec™ Real-Time 96-Well PCR Platesc

Fluorescence intensity up to ten times higher than with frosted wells

Manufacturer: Eppendorf™ 0030132734

 View more versions of this product

Catalog No. 14-281-211

  Check Availability
Add to Cart

    Specifications

    Autoclavable Yes
    Barcode Yes
    For Use With (Application) RT-PCR applications
    Material Polypropylene,Polypropylene
    Rimmed Yes
    Skirt Style Skirted
    Sterile Yes,Yes
    Well Color White
    Color White
    Product Type RT-PCR Plate
    Applications RT-PCR applications

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit