Learn More
Eppendorf™ Vacufuge™ Rotors
Fixed-angle rotors for vacuum concentrator Vacufuge plus
Manufacturer:  Eppendorf™ 022822217
 View more versions of this product
Catalog No. 05-400-248
Add to cart
Description
Description
  • For use with Model 5301
  • Variety of sizes available
Specifications
Specifications
Rotor
240 x g
5.5 cm
Vacufuge Plus Vacuum Concentrators
12 x 20 mL
1,400 rpm
3.1 cm
Fixed