Eppendorf™ Varispenser™ plus/Varispenser Bottle-Top Reagent Dispensers Available on GSA/VA Contract for Federal Government customers only.

Bottle-top reagent dispenser for taking aliquots of liquid from large supply bottles

Manufacturer: Eppendorf™ 4960000035

 View more versions of this product

Catalog No. 13-687-262

  Check Availability
Add to Cart

    Specifications

    Warranty 2 Years
    Volume Range 2 to 10mL
    Dispensing Interval 0.2mL
    Product Type Bottletop Reagent Dispenser
    Includes Telescopic tube, 3 adapters for 28, 40, 45mm O.D. bottle thread
    Autoclavable Yes
    Accuracy ±0.5%
    Precision ≤0.1%
    Certifications/Compliance EN ISO 8655

    • "Purchase two Eppendorf™ Varispenser™ or Varispenser Plus Bottletop Dispensers and get a third at no additional cost. To take advantage of this offer, apply for the NLSU program.

      New Offer Buy Now

      Expires 2018-06-30 Redemption Only!

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit