Learn More
Eppendorf™ Varispenser™ plus/Varispenser Bottle-Top Reagent Dispensers Available on GSA/VA Contract for Federal Government customers only.
Bottle-top reagent dispenser for taking aliquots of liquid from large supply bottles
Manufacturer:  Eppendorf™ 4961000063
 View more versions of this product
Catalog No. 13-687-271
  Check Availability
Add to cart
Specifications
Description
  • PFA-sealing of the slide piston prevents jamming Fine adjustment depending on viscosity of liquid No crystallization of liquid
  • No dripping when valve closed Telescopic tube for use with high bottles Rapid volume setting using precise graduation scale Patented safety valve for deairing without loss of reagent Cannel-sealing cap prevents media contact Autoclavable
  • Extremely chemical-resistant
  • Varispenser plus features safety valve and fine adjustment
Description & Specifications
Specifications
20 to 100mL
±0.5%
EN ISO 8655
Bottletop Reagent Dispenser
2 Years
Yes
≤0.1%
2mL
Telescopic tube, 3 adapters for 32, 38, 40mm O.D. bottle thread, recirculation valve, valve toggle and media specific fine adjustment
Promotions
Promotions
  • Eppendorf North America
    "Purchase two Eppendorf™ Varispenser™ or Varispenser Plus Bottletop Dispensers and get a third at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.
    06-30-2018