Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Eprosartan mesylate, Tocris™
Potent and selective AT1 receptor antagonist
Manufacturer: R&D Systems 4696/10
Quantity 10mg
 View more versions of this product
Catalog No. 46-961-0
  N/A
Add to Cart
Related Products
Product Content Feedback