Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EPS15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117938
This item is not returnable.
View return policy
Description
EPS15 Polyclonal Antibody for Western Blot, IHC (P), IP
EPS15 is a ubiquitously expressed protein that is involved in cell growth regulation and may be involved in the regulation of mitogenic signals and control of cell proliferation. Phosphorylation of EPS15 on Tyrosine 849 upon DNA damage is involved in the internalization of EGFR. A chromosomal aberration involving EPS15 is also found in acute leukemias, resulting in a rogue activator protein.
Specifications
EPS15 | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
P42566 | |
EPS15 | |
E. coli-derived Human EPS15 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
EPS15 | |
2410112D09Rik; AF1P; AF-1P; AF-1p protein; ALL1 fused gene from chromosome 1; epidermal growth factor pathway substrate 15; epidermal growth factor receptor pathway substrate 15; epidermal growth factor receptor substrate 15; EPS15; MLLT5; protein AF-1p; Protein Eps15; RGD1307641 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
2060 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction