Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPS15R Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | EPS15R |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
EPS15R Polyclonal specifically detects EPS15R in Human samples. It is validated for Western Blot.Specifications
EPS15R | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
58513 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
epidermal growth factor receptor pathway substrate 15-like 1, epidermal growth factor receptor substrate 15-like 1, epidermal growth factor receptor substrate EPS15R, EPS15R, Eps15-related protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 475-754 of human EPS15R (NP_001245304.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title