Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eps8 Mouse anti-Human, Alexa Fluor™ 532, Clone: 18F5B11, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227344AF532
Description
Eps8 Monoclonal specifically detects Eps8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Eps8 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
EPS8 | |
Amino acids 701-882 of human EPS8 protein were used as the immunogen for the EPS8 antibody. | |
Primary | |
Human | |
IgG3 κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
18F5B11 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
epidermal growth factor receptor kinase substrate 8, epidermal growth factor receptor pathway substrate 8 | |
Mouse | |
0.1 mL | |
2059 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title