Learn More
Eps8 Mouse anti-Human, Alexa Fluor™ 594, Clone: 18F5B11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227344AF594
Catalog No. NP227344A94
Add to cart
Description
Description
Eps8 Monoclonal antibody specifically detects Eps8 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Eps8
18F5B11
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
epidermal growth factor receptor kinase substrate 8, epidermal growth factor receptor pathway substrate 8
Mouse
IgG3 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
EPS8
Amino acids 701-882 of human EPS8 protein were used as the immunogen for the EPS8 antibody.
0.1 mL
Primary
2059