Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPS8L1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156689
Description
EPS8L1 Polyclonal specifically detects EPS8L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EPS8L1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
DRC3EPS8R1, epidermal growth factor receptor kinase substrate 8-like protein 1, Epidermal growth factor receptor pathway substrate 8-related protein 1, EPS8-like 1, EPS8-like protein 1, EPS8-related protein 1, FLJ20258, MGC23164, MGC4642 | |
Rabbit | |
Protein A purified | |
RUO | |
54869 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q8TE68-2 | |
EPS8L1 | |
Synthetic peptides corresponding to EPS8L1(EPS8-like 1) The peptide sequence was selected from the N terminal of EPS8L1. Peptide sequence QRDRSPAAETPPLQRRPSVRAVISTVERGAGRGRPQAKPIPEAEEAQRPE. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Equine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Canine: 85%. | |
Human, Mouse, Rat, Canine, Equine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction