Learn More
Abnova™ EPS8L2 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00064787T01
Catalog No. 89-943-496
Add to cart
Specifications
Specifications
Western Blot
293T
NM_022772.2
pCMV-EPS8L2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
EPS8L2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 78.76kDa
Transient overexpression cell lysate was tested with anti-EPS8L2 antibody (Mfr. No. H00064787-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)