Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epsilon 1 Tubulin Mouse anti-Human, Allophycocyanin, Clone: 7G3B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237244APC

Catalog No. NBP237244T

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Epsilon 1 Tubulin
7G3B2
Flow Cytometry
Purified
TUBE1
Purified recombinant fragment of human TUBE1 (AA: 314-472) expressed in E. Coli.
Protein A or G purified
Apoptosis, Cancer, Cell Cycle and Replication, Cytoskeleton Markers, Tumor Suppressors
Primary
51175
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
dJ142L7.2, Epsilon-tubulin, FLJ22589, FLJ44203, TUBEepsilon-tubulin, tubulin epsilon chain, tubulin, epsilon 1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit