Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epsin-2 Mouse anti-Human, Monkey, PE-Cyanine7, Clone: OTI1G3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271701PECY7

Catalog No. N271701PEC7


Add to cart

Description

Description

Epsin-2 Monoclonal antibody specifically detects Epsin-2 in Human, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

Epsin-2
OTI1G3
Flow Cytometry
Affinity Purified, Conjugated
EPN2
Full length recombinant protein of human Epsin-2 (NP_055779) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Monkey
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
Eps15 binding protein, EPS-15-interacting protein 2, epsin 2, epsin-2, KIAA1065EHB21
Mouse
IgG1
0.1 mL
Primary
22905.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit