Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epsin-2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | Epsin-2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Epsin-2 Polyclonal specifically detects Epsin-2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Epsin-2 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
22905 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Eps15 binding protein, EPS-15-interacting protein 2, epsin 2, epsin-2, KIAA1065EHB21 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of mouse Epsin-2 (XP_006532226.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title