Learn More
Epstein-Barr virus EA-R Mouse anti-Epstein-Barr virus, Clone: EA072, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13541
Catalog No. 89-033-504
Add to cart
Description
Description
Mouse monoclonal antibody raised against Epstein-Barr virus (EBV) EA-R.
  • High specificity
  • High reactivity
Specifications
Specifications
Epstein-Barr virus EA-R
EA072
Mouse monoclonal antibody raised against Epstein-Barr virus (EBV) EA-R.
In PBS, pH 7.4.
Mouse
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Epstein-Barr virus
ELISA, Immunofluorescence, Western Blot
Unconjugated
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
IgG1, kappa
RUO
Primary
This antibody reacts with EA-R of Epstein-Barr virus.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)