Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epstein Barr Virus Mouse anti-Virus, Clone: CS1, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Epstein Barr Virus |
---|---|
Clone | CS1 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, ELISA |
Applications | Western Blot, ELISA |
Description
Epstein Barr Virus Monoclonal specifically detects Epstein Barr Virus in Virus samples. It is validated for Western Blot, ELISA.Specifications
Epstein Barr Virus | |
0.2 mg/ml | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Virus | |
Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1 | |
IgG1 κ | |
Protein A or G purified |
CS1 | |
Western Blot 1-2 ug/ml, ELISA | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
Recombinant fusion protein containing the sequence of bacterial beta-galactosidase and the carboxyl half of EBV-encoded LMP | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title