Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epstein Barr Virus Mouse anti-Virus, Clone: CS3, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen Epstein Barr Virus
Clone CS3
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA
Applications Western Blot, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122736
View Documents
Novus Biologicals
NBP30763120UG
20 μg
Each for $138.37
N/A
Due to product restrictions, please to purchase or view availability for this product.
NB122735
View Documents
Novus Biologicals
NBP30763110UG
100 μg
Each for $352.89
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Epstein Barr Virus Monoclonal antibody specifically detects Epstein Barr Virus in Virus samples. It is validated for Western Blot, ELISA
Specifications

Specifications

Epstein Barr Virus
0.2 mg/ml
Western Blot, ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
Recombinant fusion protein containing the sequence of bacterial beta-galactosidase and the carboxyl half of EBV-encoded LMP
Monoclonal
Protein A or G purified
CS3
Western Blot 1-2 ug/ml, ELISA
Unconjugated
Mouse
Virus
Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit