Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epstein-Barr virus p18 Goat anti-Epstein-Barr virus, Polyclonal, Abnova™

Goat Polyclonal Antibody

Manufacturer:  Abnova Corporation PAB29841

Catalog No. 89-033-686


Add to cart

Description

Description

Goat polyclonal antibody raised against Epstein-Barr virus (EBV) p18.

  • High specificity
  • High reactivity
Specifications

Specifications

Epstein-Barr virus p18
Unconjugated
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
Recombinant Epstein-Barr virus viral-capsid antigen (EBV-VCA) p18.
RUO
Primary
Epstein-Barr virus
ELISA, Immunofluorescence, Western Blot
Goat polyclonal antibody raised against Epstein-Barr virus (EBV) p18.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Goat
500 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)