Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EpsteinBarrVirusNuclearAntigen3A Sheep anti-Virus, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP266589

Catalog No. NBP266589


Add to cart

Description

Description

EpsteinBarrVirusNuclearAntigen3A Polyclonal antibody specifically detects EpsteinBarrVirusNuclearAntigen3A in Virus samples. It is validated for Western Blot.
Specifications

Specifications

EpsteinBarrVirusNuclearAntigen3A
Unconjugated
PBS with 0.08% Sodium Azide
Sheep
IgG
250μg
Primary
Virus
Western Blot
Western Blot 1:100-1:2000
Purified
Recombinant EBNA-3A protein
Ammonium sulfate precipitation
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit