Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EpsteinBarrVirusNuclearAntigen6 Sheep anti-Virus, Polyclonal, Novus Biologicals
Sheep Polyclonal Antibody
Manufacturer: Novus Biologicals NBP266598
Description
EpsteinBarrVirusNuclearAntigen6 Polyclonal antibody specifically detects EpsteinBarrVirusNuclearAntigen6 in Virus samples. It is validated for Western Blot.Specifications
EpsteinBarrVirusNuclearAntigen6 | |
Unconjugated | |
PBS with 0.08% Sodium Azide | |
Epstein Barr Virus Nuclear Antigen 3C | |
Recombinant EBNA-3C protein | |
Ammonium sulfate precipitation | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Polyclonal |
Western Blot | |
Western Blot 1:100-1:2000 | |
Purified | |
Sheep | |
IgG | |
250μg | |
Primary | |
Virus |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title