Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPX Mouse anti-Human, Clone: EPO104, Novus Biologicals™
Mouse Monoclonal Antibody
$229.00 - $459.00
Specifications
Antigen | EPX |
---|---|
Clone | EPO104 |
Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml, SDS-Page |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), SDS-Page |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP232844A
|
Novus Biologicals
NBP2328440.1MG |
0.1 mg |
Each for $459.00
|
|
NBP232844F
|
Novus Biologicals
NBP2328440.02MG |
0.02 mg |
Each for $229.00
|
|
NBP232844B
|
Novus Biologicals
NBP2328440.2MG |
0.2 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
EPX Monoclonal specifically detects EPX in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
EPX | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
EC 1.11.1.7, eosinophil peroxidase, EPOEPER, EPPEC 1.11.1, EPX-PEN | |
EPX | |
IgG1 κ | |
Protein A purified | |
Peripheral blood granulocytes are classified into neutrophils, basophils and eosinophils according to the staining characteristics of their cytoplasmic granules. Granule proteins are released by physiologic and pharmacologic stimuli and play important roles in both normal and pathological host immune responses. Eosinophil major basic protein and eosinophil peroxidase (EPX) are granule proteins specific to the eosinophil. AHE-1 recognizes human EPX, a granule protein specific to eosinophils. It does not cross-react with eosinophil major basic protein, elastase, cathepsin G, esterase N, thrombin, plasmin, kallikrein, lactoferrin, or transferrin. This MAb stains eosinophils only and does not stain other peripheral blood cells, including platelets, neutrophils, monocytes, lymphocytes or red blood cells. Human EPX gene product can form a tetramer of two light chains and two heavy chains. Other peroxidase family members include myeloperoxidase (MPO), lactoperoxidase (LPO), and thyroid peroxidase (TPO). |
EPO104 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), SDS-Page | |
Unconjugated | |
Mouse | |
Human | |
P11678 | |
8288 | |
Human eosinophils from a patient with hypereosinophilic syndrome | |
Primary | |
Store at 4C. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title