Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPX Rabbit anti-Human, Clone: EPX/3908R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen EPX
Clone EPX/3908R
Concentration 0.2 mg/ml
Dilution Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122251
View Documents
Novus Biologicals
NBP30738910UG
100 μg
Each for $406.52
Add to cart
 
NB122252
View Documents
Novus Biologicals
NBP30738920UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

EPX Monoclonal antibody specifically detects EPX in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

EPX
0.2 mg/ml
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
8288
IgG
Primary
Store at 4°C.
EPX/3908R
Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin
Unconjugated
Rabbit
Human
EC 1.11.1.7, eosinophil peroxidase, EPOEPER, EPPEC 1.11.1, EPX-PEN
Human eosinophils from a patient with hypereosinophilic syndrome
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit