Learn More
Azzota Corp EPZ-015666 DFS Item
Manufacturer:  Azzota Corp C1132
EPZ-015666 (GSK3235025) is a potent and selective and orally active PRMT5 inhibitor with a half-maximal inhibitory concentration (IC50) of 22 nM and broad selectivity against a panel of other histone methyltransferases. Please request if need more quantities
Catalog No. 50-161-3102