Learn More
Azzota Corp EPZ-031686 DFS Item
Manufacturer:  Azzota Corp C1404
EPZ-031686 is a noncompetitive inhibitor for SMYD3 and MEKK2 with a Ki=1.2 and 1.1 nM respectively. In vivo, EPZ031686 shows good bioavailability following oral dosing in mice. Please request if need more quantities
Catalog No. 50-161-3212