Learn More
Equine IgG, F(ab)2, HRP, Isotype control, Novus Biologicals™
Horse Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197159
Catalog No. NBP197159
Add to cart
Description
Description
IgG F(ab)2 antibody specifically detects IgG F(ab)2 in Equine samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
IgG F(ab)2
HRP
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Gentamicin Sulfate
Equine
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
LYOPH
Restore with 1.0mL of deionized water (or equivalent).
Conjugated
IgG2
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Equine IgG, anti-Equine IgG F(ab)2 and anti-Equine Serum. No reaction was observed against anti-Equine IgG F(c) or anti-Pepsin.
Equine