Learn More
Raybiotech Inc Equine IL-2 ELISA Kit
SDP
Supplier:  Raybiotech Inc ELEIL21
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Equine IL-2. Other species not determined.
Catalog No. 50-194-8907