Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Equine Rotavirus Antigen ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of horse rotavirus antigen.

$602.00

Specifications

Species Equine
Sample Type feces
Research Discipline Cell Biology
Regulatory Status RUO
Sample Volume 50 to 100μL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601321KT View Documents Novus Biologicals™
NBP2601321KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Equine Rotavirus Antigen ELISA Kit (Colorimetric) measures Equine Rotavirus Antigen in feces.
Specifications

Specifications

Equine
Cell Biology
50 to 100μL
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
Instruction manual, Positive Control, Negative Control, Stop Solution, Wash Buffer, Substrate A, HRP-conjugate, Assay plate, Adhesive Strip (For 96 wells)
ELISA Kit (Colorimetric)
feces
RUO
For detection of horse rotavirus antigen
Rotavirus Antigen
ELISA
Qualitative

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit