Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Equine Vitamin A ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of Vitamin A.

$732.50

Specifications

Species Equine
Sensitivity 1.402ng/mL
Sample Type Serum, plasma
Research Discipline Lipid and Metabolism
Regulatory Status RUO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601891KT View Documents Novus Biologicals™
NBP2601891KIT
1 Kit Each for $732.50
Add to cart
 
Description

Description

The Equine Vitamin A ELISA Kit (Colorimetric) measures equine Vitamin A in serum, plasma.
Specifications

Specifications

Equine
Serum, plasma
RUO
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA
1.875 to 30ng/mL
1.402ng/mL
Lipid and Metabolism
50 to 100μL
Vitamin A
ELISA Kit (Colorimetric)

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit