Learn More
E.R. Advanced Ceramics Jars for Jar Mills
Manufacturer:  E.R. Advanced Ceramics J7740E
 View more versions of this product
Catalog No. 08-382C
Add to cart
Description
Description
For 08-381 Series Jar Mills.
  • Porcelain fortified with alumina
  • Strong and chemical resistant
  • Extra-wide mouth
  • Locking lid with bar and hand wheel
  • Neoprene gasket
  • Precision-ground cover and gasket seal
Specifications
Specifications
64.24 oz.
64.24 oz., 1900mL
6.25 in.
15.87cm
22.22cm
Fortified Porcelain, Alumina
3.17kg
1900mL
2.75 in.
6.98cm
8.75 in.
Locking lid with bar, hand wheel, Neoprene gasket, precision ground cover, gasket seal
7 lb.
08-381 Series jar mill