Learn More
E.R. Advanced Ceramics Jars for Jar Mills
Manufacturer:  E.R. Advanced Ceramics J7742E
 View more versions of this product
Catalog No. 08-382E
Add to cart
Description
Description
For 08-381 Series Jar Mills.
  • Porcelain fortified with alumina
  • Strong and chemical resistant
  • Extra-wide mouth
  • Locking lid with bar and hand wheel
  • Neoprene gasket
  • Precision-ground cover and gasket seal
Specifications
Specifications
280.65 oz.
280.65 oz., 8300mL
27.94cm
32.38cm
Fortified Porcelain, Alumina
8300mL
11 in.
12.75 in.
Locking lid with bar, hand wheel, Neoprene gasket, precision ground cover, gasket seal
08-381 Series jar mill