Learn More
E.R. Advanced Ceramics Jars for Jar Mills
Manufacturer:  E.R. Advanced Ceramics J7743E
 View more versions of this product
Catalog No. 08-382F
Add to cart
Description
Description
For 08-381 Series Jar Mills.
  • Porcelain fortified with alumina
  • Strong and chemical resistant
  • Extra-wide mouth
  • Locking lid with bar and hand wheel
  • Neoprene gasket
  • Precision-ground cover and gasket seal
Specifications
Specifications
422.67 oz.
422.67 oz., 12500mL
33.02cm
33.02cm
Fortified Porcelain, Alumina
12500mL
13 in.
13 in.
Locking lid with bar, hand wheel, Neoprene gasket, precision ground cover, gasket seal
08-381 Series jar mill