Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ER alpha/NR3A1 Mouse anti-Human, Alexa Fluor™ 350, Clone: ER503, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP233323AF350

Catalog No. NP233323A30


Add to cart

Description

Description

ER alpha/NR3A1 Monoclonal antibody specifically detects ER alpha/NR3A1 in Human samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications

Specifications

ER alpha/NR3A1
ER503
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3&asterisk;4,56,7&asterisk;/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4&asterisk;5,6,7&asterisk;/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
ESR1
Recombinant human Estrogen Receptor alpha protein (aa2-185)
0.1 mL
Primary
2099
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit