Learn More
ER alpha/NR3A1 Mouse anti-Human, Clone: C.15200009, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259201
Catalog No. NBP259201
Add to cart
Description
Description
ER alpha/NR3A1 Monoclonal antibody specifically detects ER alpha/NR3A1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry.
Specifications
Specifications
ER alpha/NR3A1
C.15200009
Western Blot 7 ug/ml, Chromatin Immunoprecipitation 5 ug, Immunohistochemistry 15 ug/ml, Immunocytochemistry
Purified
ESR1
The exact immunogen is propietary information.
ammonium sulphate purified
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry
Unconjugated
PBS with 0.05% Sodium Azide
ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1
Mouse
IgG3
50 ug
Angiogenesis, Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary
2099